missing translation for 'onlineSavingsMsg'
Learn More

Tri-p-tolylsulfonium Trifluoromethanesulfonate 95.0+%, TCI America™

Catalog No. T20421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20421G 1 g
T20425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20421G Supplier TCI America Supplier No. T20421G

Chemical Identifiers

CAS 127820-38-6
Molecular Formula C22H21F3O3S2
Molecular Weight (g/mol) 454.52
MDL Number MFCD06797188
InChI Key ZZJNLOGMYQURDL-UHFFFAOYSA-M
Synonym Tris(4-methylphenyl)sulfonium Trifluoromethanesulfonate, Tri-p-tolylsulfonium Triflate
PubChem CID 21863653
IUPAC Name tris(4-methylphenyl)sulfanium trifluoromethanesulfonate
SMILES [O-]S(=O)(=O)C(F)(F)F.CC1=CC=C(C=C1)[S+](C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Melting Point 106°C
Color White
Quantity 1 g
Formula Weight 454.52
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tri-p-tolylsulfonium Trifluoromethanesulfonate