missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-p-tolylamine 98.0+%, TCI America™
Supplier: TCI America T255825G
Specifications
| Tri-p-tolylamine | |
| 117°C | |
| 170°C | |
| MFCD00674043 | |
| tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamine | |
| CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C | |
| 287.406 | |
| 287.41 | |
| Crystalline Powder |
| 1159-53-1 | |
| White-Yellow | |
| C21H21N | |
| 25 g | |
| YXYUIABODWXVIK-UHFFFAOYSA-N | |
| 4-methyl-N,N-bis(4-methylphenyl)aniline | |
| 70873 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1159-53-1 | |
| 287.406 | |
| YXYUIABODWXVIK-UHFFFAOYSA-N | |
| 70873 | |
| CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C |
| C21H21N | |
| MFCD00674043 | |
| tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamine | |
| 4-methyl-N,N-bis(4-methylphenyl)aniline |
Safety and Handling
TSCA : Yes