missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-p-cresyl Phosphate 98.0+%, TCI America™
$119.36 - $411.98
Chemical Identifiers
| CAS | 78-32-0 |
|---|---|
| Molecular Formula | C21H21O4P |
| Molecular Weight (g/mol) | 368.37 |
| MDL Number | MFCD00041908 |
| InChI Key | BOSMZFBHAYFUBJ-UHFFFAOYSA-N |
| Synonym | tri-p-cresyl phosphate, tri-p-tolyl phosphate, tris 4-methylphenyl phosphate, tpcp, phosphoric acid, tris 4-methylphenyl ester, phosphoric acid, tri-p-tolyl ester, phosphoric acid, tri 4-tolyl ester, phosphoric acid, tris methylphenyl ester, p-tolyl phosphate c7h7o 3po, tris-p-tolyl phosphate |
| PubChem CID | 6529 |
| IUPAC Name | tris(4-methylphenyl) phosphate |
| SMILES | CC1=CC=C(OP(=O)(OC2=CC=C(C)C=C2)OC2=CC=C(C)C=C2)C=C1 |
Chemical Identifiers
| 78-32-0 | |
| 368.37 | |
| BOSMZFBHAYFUBJ-UHFFFAOYSA-N | |
| 6529 | |
| CC1=CC=C(OP(=O)(OC2=CC=C(C)C=C2)OC2=CC=C(C)C=C2)C=C1 |
| C21H21O4P | |
| MFCD00041908 | |
| tri-p-cresyl phosphate, tri-p-tolyl phosphate, tris 4-methylphenyl phosphate, tpcp, phosphoric acid, tris 4-methylphenyl ester, phosphoric acid, tri-p-tolyl ester, phosphoric acid, tri 4-tolyl ester, phosphoric acid, tris methylphenyl ester, p-tolyl phosphate c7h7o 3po, tris-p-tolyl phosphate | |
| tris(4-methylphenyl) phosphate |
Specifications
| 78-32-0 | |
| White-Yellow | |
| C21H21O4P | |
| 5 g | |
| tri-p-cresyl phosphate, tri-p-tolyl phosphate, tris 4-methylphenyl phosphate, tpcp, phosphoric acid, tris 4-methylphenyl ester, phosphoric acid, tri-p-tolyl ester, phosphoric acid, tri 4-tolyl ester, phosphoric acid, tris methylphenyl ester, p-tolyl phosphate c7h7o 3po, tris-p-tolyl phosphate | |
| CC1=CC=C(OP(=O)(OC2=CC=C(C)C=C2)OC2=CC=C(C)C=C2)C=C1 | |
| 368.37 | |
| 368.37 | |
| Crystalline Powder |
| 77°C | |
| 430°C | |
| MFCD00041908 | |
| 3077 | |
| BOSMZFBHAYFUBJ-UHFFFAOYSA-N | |
| tris(4-methylphenyl) phosphate | |
| 6529 | |
| ≥98.0% (GC) | |
| Tri-p-cresyl Phosphate |
Safety and Handling
EINECSNumber : (3)-2522&(3)-2613&(3)-3363
RTECSNumber : TC9330000
TSCA : Yes