missing translation for 'onlineSavingsMsg'
Learn More

Tri-p-cresyl Phosphate 98.0+%, TCI America™

Catalog No. T22095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22095G 5 g
T220925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22095G Supplier TCI America Supplier No. T22095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-32-0
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.37
MDL Number MFCD00041908
InChI Key BOSMZFBHAYFUBJ-UHFFFAOYSA-N
Synonym tri-p-cresyl phosphate, tri-p-tolyl phosphate, tris 4-methylphenyl phosphate, tpcp, phosphoric acid, tris 4-methylphenyl ester, phosphoric acid, tri-p-tolyl ester, phosphoric acid, tri 4-tolyl ester, phosphoric acid, tris methylphenyl ester, p-tolyl phosphate c7h7o 3po, tris-p-tolyl phosphate
PubChem CID 6529
IUPAC Name tris(4-methylphenyl) phosphate
SMILES CC1=CC=C(OP(=O)(OC2=CC=C(C)C=C2)OC2=CC=C(C)C=C2)C=C1

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 430°C
Quantity 5 g
UN Number 3077
Formula Weight 368.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri-p-cresyl Phosphate
EINECSNumber (3)-2522&(3)-2613&(3)-3363
RTECSNumber TC9330000
TSCA Yes