missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-o-tolylbismuthine 98.0+%, TCI America™
Supplier: TCI America T19705G
Specifications
| Tri-o-tolylbismuthine | |
| 134°C | |
| C21H21Bi | |
| 5 g | |
| DWIMJCTYUMGECA-UHFFFAOYSA-N | |
| tris(2-methylphenyl)bismuthane | |
| 10972889 | |
| ≥98.0% (HPLC,T) |
| 10050-08-5 | |
| White | |
| MFCD00123270 | |
| Tris(2-methylphenyl)bismuthine | |
| CC1=CC=CC=C1[Bi](C2=CC=CC=C2C)C3=CC=CC=C3C | |
| 482.379 | |
| 482.38 | |
| Crystalline Powder |
Chemical Identifiers
| 10050-08-5 | |
| 482.379 | |
| DWIMJCTYUMGECA-UHFFFAOYSA-N | |
| 10972889 | |
| CC1=CC=CC=C1[Bi](C2=CC=CC=C2C)C3=CC=CC=C3C |
| C21H21Bi | |
| MFCD00123270 | |
| Tris(2-methylphenyl)bismuthine | |
| tris(2-methylphenyl)bismuthane |
Safety and Handling
TSCA : No