missing translation for 'onlineSavingsMsg'
Learn More

Tri-o-tolyl phosphate, 96%

Catalog No. p-2692406
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC366460250 25 g Glass bottle
AC366461000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC366460250 Supplier Thermo Scientific Chemicals Supplier No. 366460250

CAS: 78-30-8 | C21H21O4P | 368.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-30-8
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.37
MDL Number MFCD00041906
InChI Key YSMRWXYRXBRSND-UHFFFAOYSA-N
Synonym tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
PubChem CID 6527
IUPAC Name tris(2-methylphenyl) phosphate
SMILES CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C

Specifications

Melting Point -25°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 410°C
Flash Point 225°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.561
Quantity 25 g
Specific Gravity 1.2
Merck Index 15, 9941
Solubility Information Solubility in water: 0.01g/100mL (25°C). Other solubilities: slightly soluble in alcohol,soluble in ether
Formula Weight 368.36
Percent Purity 96%
Physical Form Viscous Liquid
Chemical Name or Material Tri-o-tolyl phosphate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes damage to organs.
Precautionary Statement GHS P Statement:
IF exposed: Call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-103-5

RUO – Research Use Only