missing translation for 'onlineSavingsMsg'
Learn More

Tri-O-acetyl-D-glucal 96.0+%, TCI America™

Catalog No. t159625g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T159625G 25 g
T15965G 5 g
2 options

Catalog No. T159625G

Supplier: TCI America T159625G

Only null left

Chemical Identifiers

CAS 2873-29-2
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.253
MDL Number MFCD00063253
InChI Key LLPWGHLVUPBSLP-UTUOFQBUSA-N
Synonym tri-o-acetyl-d-glucal, 3,4,6-tri-o-acetyl-d-glucal, 2r,3s,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-glucal triacetate, 3,4,6-tri-o-acetyl-1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, triacetate, tri-o-acetylglucal, triacetyl-d-glucal, 2r,3s,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, tri-o-acetyl-glucal
PubChem CID 688303
IUPAC Name [(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C

Specifications

Melting Point 55°C
Color White
Quantity 25 g
Formula Weight 272.25
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri-O-acetyl-D-glucal