missing translation for 'onlineSavingsMsg'
Learn More

Tri-O-acetyl-D-glucal, 99%

Catalog No. AC139240250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139240250 25 g Glass bottle
AC139241000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC139240250 Supplier Thermo Scientific Chemicals Supplier No. 139240250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2873-29-2 | C12H16O7 | 272.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2873-29-2
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.25
MDL Number MFCD00063253
InChI Key LLPWGHLVUPBSLP-UTUOFQBUSA-N
Synonym tri-o-acetyl-d-glucal, 3,4,6-tri-o-acetyl-d-glucal, 2r,3s,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-glucal triacetate, 3,4,6-tri-o-acetyl-1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, triacetate, tri-o-acetylglucal, triacetyl-d-glucal, 2r,3s,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, tri-o-acetyl-glucal
PubChem CID 688303
IUPAC Name [(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C

Specifications

Melting Point 51.0°C to 53.0°C
Color White
Flash Point >110°C
Loss on Drying 0.5% max. (30°C, 15 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 17, I, 111
Specific Rotation Condition −15° (20°C c=2,EtOH)
Specific Rotation − 15.00
Solubility Information Solubility in water: slightly soluble
Formula Weight 272.25
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tri-O-acetyl-D-glucal, 99%
Show More Show Less
EINECSNumber 220-709-0

RUO – Research Use Only