missing translation for 'onlineSavingsMsg'
Learn More

Tri-O-acetyl-D-galactal 95.0+%, TCI America™

Catalog No. T17341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17341G 1 g
T17345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17341G Supplier TCI America Supplier No. T17341G

Chemical Identifiers

CAS 4098-06-0
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.25
MDL Number MFCD00064092
InChI Key LLPWGHLVUPBSLP-UHFFFAOYNA-N
Synonym 3,4,6-tri-o-acetyl-d-galactal, tri-o-acetyl-d-galactal, 2r,3r,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-arabino-hex-5-enitol, 2,6-anhydro-5-deoxy-, triacetate, 1,3,4-tri-o-acetyl-2,6-anhydro-5-deoxyhex-5-enitol, d-galactal triacetate, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol triacetate, 2r,3r,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, galactal triacetate, tri-o-acetylgalactal
PubChem CID 640125
IUPAC Name [3,4-bis(acetyloxy)-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OCC1OC=CC(OC(C)=O)C1OC(C)=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 272.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tri-O-acetyl-D-galactal
TSCA No
Recommended Storage Refrigerator