missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylphosphine, tech. 90%

Catalog No. AAL15744AP
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15744AP 500 mL
AAL15744AC 25 mL
AAL15744AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL15744AP Supplier Thermo Scientific Chemicals Supplier No. L15744AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4731-53-7 | C24H51P | 370.646 g/mol

Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4731-53-7
Molecular Formula C24H51P
Molecular Weight (g/mol) 370.646
MDL Number MFCD00015298
InChI Key RMZAYIKUYWXQPB-UHFFFAOYSA-N
Synonym trioctylphosphine, tri-n-octylphosphine, phosphine, trioctyl, tri-n-octyl phosphine, tri-octylphosphine, trioctyl phosphine, tri degrees ctylphosphine, acmc-209k8q
PubChem CID 20851
IUPAC Name trioctylphosphane
SMILES CCCCCCCCP(CCCCCCCC)CCCCCCCC

Specifications

Density 0.831
Boiling Point 240°C (10 mmHg)
Flash Point 147°C (296°F)
Odor Garlic-like
Refractive Index 1.468
Quantity 500 mL
UN Number UN1760
Beilstein 1776995
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 370.65
Percent Purity 90%
Grade Technical
Chemical Name or Material Tri-n-octylphosphine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 225-234-2
RTECSNumber SZ3450000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only