missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylphosphine 85.0+%, TCI America™

Catalog No. p-7136314
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T050325ML 25 mL
T0503100ML 100 mL
T0503500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T050325ML Supplier TCI America Supplier No. T050325ML
Only null left

Chemical Identifiers

CAS 4731-53-7
Molecular Formula C24H51P
Molecular Weight (g/mol) 370.646
MDL Number MFCD00015298
InChI Key RMZAYIKUYWXQPB-UHFFFAOYSA-N
Synonym trioctylphosphine, tri-n-octylphosphine, phosphine, trioctyl, tri-n-octyl phosphine, tri-octylphosphine, trioctyl phosphine, tri degrees ctylphosphine, acmc-209k8q
PubChem CID 20851
IUPAC Name trioctylphosphane
SMILES CCCCCCCCP(CCCCCCCC)CCCCCCCC

Specifications

Color Yellow
Boiling Point 234°C
Quantity 25 mL
UN Number 1760
Formula Weight 370.65
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Tri-n-octylphosphine
EINECSNumber (2)-3735
RTECSNumber SZ3450000
TSCA Yes