missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-octylphosphine Oxide 95.0+%, TCI America™
Supplier: TCI America T0504100G
Specifications
| Tri-n-octylphosphine Oxide | |
| 54°C | |
| 238°C | |
| MFCD00002083 | |
| trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg | |
| CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC | |
| 386.645 | |
| 386.64 | |
| Crystalline Powder |
| 78-50-2 | |
| White | |
| C24H51OP | |
| 100 g | |
| ZMBHCYHQLYEYDV-UHFFFAOYSA-N | |
| 1-dioctylphosphoryloctane | |
| 65577 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 78-50-2 | |
| 386.645 | |
| ZMBHCYHQLYEYDV-UHFFFAOYSA-N | |
| 65577 | |
| CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC |
| C24H51OP | |
| MFCD00002083 | |
| trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg | |
| 1-dioctylphosphoryloctane |
Safety and Handling
EINECSNumber : (2)-2937
RTECSNumber : SZ1662500
TSCA : Yes