missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylphosphine Oxide 95.0+%, TCI America™
GREENER_CHOICE

Catalog No. T050425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T050425G 25 g
T0504100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T050425G Supplier TCI America Supplier No. T050425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78-50-2
Molecular Formula C24H51OP
Molecular Weight (g/mol) 386.645
MDL Number MFCD00002083
InChI Key ZMBHCYHQLYEYDV-UHFFFAOYSA-N
Synonym trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
PubChem CID 65577
IUPAC Name 1-dioctylphosphoryloctane
SMILES CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 54°C
Color White
Boiling Point 238°C
Quantity 25 g
Formula Weight 386.64
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri-n-octylphosphine Oxide
EINECSNumber (2)-2937
RTECSNumber SZ1662500
TSCA Yes