missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylphosphine oxide, 99%

Catalog No. p-3737674
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207600250 25 g Glass bottle
AC207601000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207600250 Supplier Thermo Scientific Chemicals Supplier No. 207600250
Only null left

CAS: 78-50-2 | C24H51OP | 386.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-50-2
Molecular Formula C24H51OP
Molecular Weight (g/mol) 386.64
MDL Number MFCD00002083
InChI Key ZMBHCYHQLYEYDV-UHFFFAOYSA-N
Synonym trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
PubChem CID 65577
IUPAC Name 1-dioctylphosphoryloctane
SMILES CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 50.0°C to 55.0°C
Color White to Yellow
Boiling Point 201.0°C to 202.0°C (2.0 mmHg)
Flash Point 230°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (including isomer) (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)7]3P(O)
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in cyclohexane,methanol
Formula Weight 386.64
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material Tri-n-octylphosphine oxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protecti
EINECSNumber 201-121-3

RUO – Research Use Only