missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-octylamine 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 367°C |
| Quantity | 500 mL |
| Formula Weight | 353.68 |
| Percent Purity | ≥97.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | Tri-n-octylamine |
Safety and Handling
| EINECSNumber | (2)-0143&(2)-0139 |
| RTECSNumber | RG8225000 |
| TSCA | Yes |