missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylamine 98.0+%, TCI America™

Catalog No. I036225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I036225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I036225ML Supplier TCI America Supplier No. I036225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Organic amine used as a building block in chemical synthesis.

Chemical Identifiers

CAS 1116-76-3
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.679
MDL Number MFCD00009560
InChI Key XTAZYLNFDRKIHJ-UHFFFAOYSA-N
Synonym trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
PubChem CID 14227
IUPAC Name N,N-dioctyloctan-1-amine
SMILES CCCCCCCCN(CCCCCCCC)CCCCCCCC

Specifications

Color Yellow
Boiling Point 367°C
Quantity 25 mL
UN Number 3082
Formula Weight 353.68
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material Tri-n-octylamine [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-0143&(2)-0139
RTECSNumber RG8225000
TSCA Yes