missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylamine, 97%

Catalog No. p-4509482
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140281000 100 mL Glass bottle
AC140285000 500 mL Glass bottle
AC140280025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC140281000 Supplier Thermo Scientific Chemicals Supplier No. 140281000
Only null left

CAS: 1116-76-3 | C24H51N | 353.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1116-76-3
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.66
InChI Key XTAZYLNFDRKIHJ-UHFFFAOYSA-N
Synonym trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
PubChem CID 14227
IUPAC Name N,N-dioctyloctan-1-amine
SMILES CCCCCCCCN(CCCCCCCC)CCCCCCCC

Specifications

Melting Point -34.0°C
Color White
Density 0.8090g/mL
Boiling Point 365.0°C to 367.0°C
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4480 to 1.4500
Linear Formula [CH3(CH2)7]3N
Quantity 100 mL
Beilstein 04,196
Specific Gravity 0.809
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 353.66
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Tri-n-octylamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the envir
EINECSNumber 214-242-1

RUO – Research Use Only