Learn More
Tri-n-octylamine, 95%
CAS: 1116-76-3 | C24H51N | 353.679 g/mol
Supplier: Thermo Scientific Chemicals A150670F
Description
Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.
Solubility
Miscible with chloroform. Immiscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, carbon dioxide, acids, acid chlorides and acid anhydrides.
Specifications
| Tri-n-octylamine | |
| -39°C | |
| 164°C to 168°C (0.7 mmHg) | |
| Amine-like | |
| 1.4485 | |
| MFCD00009560 | |
| UN3082 | |
| Air Sensitive | |
| Miscible with chloroform. Immiscible with water. | |
| CCCCCCCCN(CCCCCCCC)CCCCCCCC | |
| 353.679 | |
| 353.68 |
| 1116-76-3 | |
| 0.81 | |
| 158°C (316°F) | |
| C24H51N | |
| [CH3(CH2)7]3N | |
| 2500 mL | |
| 1210618 | |
| trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08 | |
| XTAZYLNFDRKIHJ-UHFFFAOYSA-N | |
| N,N-dioctyloctan-1-amine | |
| 14227 | |
| 95% |
Chemical Identifiers
| 1116-76-3 | |
| 353.679 | |
| XTAZYLNFDRKIHJ-UHFFFAOYSA-N | |
| 14227 | |
| CCCCCCCCN(CCCCCCCC)CCCCCCCC |
| C24H51N | |
| MFCD00009560 | |
| trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08 | |
| N,N-dioctyloctan-1-amine |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 214-242-1
RTECSNumber : RG8225000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only