missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylamine, 95%

Catalog No. AAA15067AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15067AE 100 mL
AAA15067AP 500 mL
AAA150670F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA15067AE Supplier Thermo Scientific Chemicals Supplier No. A15067AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1116-76-3 | C24H51N | 353.679 g/mol

Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.

Solubility
Miscible with chloroform. Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, carbon dioxide, acids, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 1116-76-3
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.679
MDL Number MFCD00009560
InChI Key XTAZYLNFDRKIHJ-UHFFFAOYSA-N
Synonym trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
PubChem CID 14227
IUPAC Name N,N-dioctyloctan-1-amine
SMILES CCCCCCCCN(CCCCCCCC)CCCCCCCC

Specifications

Melting Point -39°C
Density 0.81
Boiling Point 164°C to 168°C (0.7 mmHg)
Flash Point 158°C (316°F)
Odor Amine-like
Refractive Index 1.4485
Linear Formula [CH3(CH2)7]3N
Quantity 100 mL
UN Number UN3082
Beilstein 1210618
Sensitivity Air Sensitive
Solubility Information Miscible with chloroform. Immiscible with water.
Formula Weight 353.68
Percent Purity 95%
Chemical Name or Material Tri-n-octylamine
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 214-242-1
RTECSNumber RG8225000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only