missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-hexylamine, 96%

Catalog No. p-7027168
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2371618 50 g
AAB2371630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2371618 Supplier Thermo Scientific Chemicals Supplier No. B2371618

CAS: 102-86-3 | C18H39N | 269.517 g/mol

Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid chloridres, acid anhydrides and chloroformates.

Chemical Identifiers

CAS 102-86-3
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.517
MDL Number MFCD00009523
InChI Key DIAIBWNEUYXDNL-UHFFFAOYSA-N
Synonym tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl
PubChem CID 66022
IUPAC Name N,N-dihexylhexan-1-amine
SMILES CCCCCCN(CCCCCC)CCCCCC

Specifications

Melting Point <-75°C
Density 0.798
Boiling Point 263°C to 265°C
Flash Point 144°C (291°F)
Refractive Index 1.4415
Quantity 50 g
UN Number UN3082
Beilstein 1755828
Solubility Information Slightly miscible with water.
Formula Weight 269.52
Percent Purity 97%
Chemical Name or Material Tri-n-hexylamine
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 203-062-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only