missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin iodide, tech. 90%, stab. with copper, Thermo Scientific™

Catalog No. p-7052808
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5591106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH5591106 Supplier Thermo Scientific Chemicals Supplier No. H5591106
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7342-47-4
Molecular Formula C12H27ISn
Molecular Weight (g/mol) 416.962
MDL Number MFCD00074996
InChI Key YHHVXVNXTMIXOL-UHFFFAOYSA-M
Synonym tributyltin iodide, iodotributylstannane, stannane, iodotributyl, iodotributyltin, stannane, tributyliodo, tri-n-butyl tin iodide, tin, tri-n-butyl-, iodide, bu3sni, tri-n-butyltin iodide, tributyl iodo stannane
PubChem CID 23765
IUPAC Name tributyl(iodo)stannane
SMILES CCCC[Sn](CCCC)(CCCC)I

Specifications

Color Colorless
Density 1.46 g/mL
Boiling Point 172°C (10 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.53
Linear Formula [CH3(CH2)3]3SnI
Quantity 5 g
UN Number UN2788
Sensitivity Light sensitive
Formula Weight 416.94
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin iodide, Stabilized with copper
Show More Show Less
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
RTECSNumber WH8580000
TSCA No
Recommended Storage Ambient temperatures