missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin hydride, 97%

Catalog No. AAA1329830
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
Catalog No. Quantity
AAA1329830 250 g
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Catalog No. AAA1329830

Supplier: Thermo Scientific Chemicals A1329830

Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 688-73-3 | C12H27Sn | 290.058 g/mol

Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.

Chemical Identifiers

CAS 688-73-3
Molecular Formula C12H27Sn
Molecular Weight (g/mol) 290.058
MDL Number MFCD00009416
InChI Key PIILXFBHQILWPS-UHFFFAOYSA-N
Synonym tributylstannanyl, tributlytin, tributylstannic hydride, tributyl tin, tri-n-butylstannane hydride, unii-4xdx163p3d, tin, tri-n-butyl-, hydride, tri-n-butylstannyl radical, stannane, tri-n-butyl-, hydride, tributyl-tin
PubChem CID 3032732
IUPAC Name tributyltin
SMILES CCCC[Sn](CCCC)CCCC

Specifications

Melting Point 0°C to 1°C
Density 1.098
Boiling Point 68°C to 74°C (0.3 mmHg)
Flash Point 40°C (104°F)
Odor Odorless
Refractive Index 1.473
Quantity 250 g
UN Number UN2929
Beilstein 3587329
Sensitivity Air sensitive; May form a precipitate
Solubility Information Immiscible with water.
Formula Weight 291.07
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin hydride
Show More Show Less
Hazard Category H301-H312-H315-H317-H319-H360FD-H372
Hazard Statement GHS H Statement
H301-H372-H226-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Flammable liquid and vapour.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 211-704-4
RTECSNumber WH8675000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only