missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylgermanium hydride, 97+%

Catalog No. p-7052156
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4268803 1 g
AA4268806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4268803 Supplier Thermo Scientific Chemicals Supplier No. 04268803
Only null left

Reduction of benzylic chlorides | CAS: 998-39-0 | C12H27Ge | 243.978 g/mol

Tri-n-butylgermanium hydride is used in the reduction of benzylic chlorides. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 998-39-0
Molecular Formula C12H27Ge
Molecular Weight (g/mol) 243.978
MDL Number MFCD00077987
InChI Key UMRPCNGKANTZSN-UHFFFAOYSA-N
Synonym tri-n-butylgermane, tributylgermylradical, tri-n-butylgermanium, hydride
PubChem CID 6327830
IUPAC Name tributylgermanium
SMILES CCCC[Ge](CCCC)CCCC

Specifications

Color Colorless to Yellow
Density 0.92 g/mL
Boiling Point 123°C (20 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.4508
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Not miscible.
Formula Weight 244.95
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Tri-n-butylgermanium hydride
Show More Show Less
Hazard Category H302+H312+H332-H318
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only