missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl(vinyl)tin, 96%

CAS: 7486-35-3 | C14H30Sn | 317.104 g/mol

$84.37 - $84.37

Chemical Identifiers

CAS 7486-35-3
Molecular Formula C14H30Sn
MDL Number MFCD00009421
InChI Key QIWRFOJWQSSRJZ-UHFFFAOYSA-N
Synonym tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
PubChem CID 81998
IUPAC Name tributyl(ethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2021503
View Documents
Thermo Scientific Chemicals
L2021503
1 g
Each for $84.37
Only null left
 
Description

Description

Tributyl(vinyl)tin is used as a reagent in the palladium catalyzed synthesis of allyl and benzyl ethers. It is also a vinyl nucleophile for bromoacetylene and bromoaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tributyl(vinyl)tin is used as a reagent in the palladium catalyzed synthesis of allyl and benzyl ethers. It is also a vinyl nucleophile for bromoacetylene and bromoaromatics.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

7486-35-3
MFCD00009421
tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
tributyl(ethenyl)stannane
C14H30Sn
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
81998
CCCC[Sn](CCCC)(CCCC)C=C

Specifications

7486-35-3
104°C to 106°C (3.5 mmHg)
C14H30Sn
MFCD00009421
UN2788
Moisture sensitive
Not miscible or difficult to mix in water.
CCCC[Sn](CCCC)(CCCC)C=C
317.104
317.11
Tri-n-butyl(vinyl)tin
1.085
>110°C (230°F)
1.478
1 g
3537662
tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
tributyl(ethenyl)stannane
81998
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

EINECSNumber : 231-291-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only