Learn More
Tri-n-butyl phosphite, 94%
CAS: 102-85-2 | C12H27O3P | 250.319 g/mol
Supplier: Thermo Scientific Chemicals L14307AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsTributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.
Solubility
Not miscible or difficult to mix in water.
Notes
Air and moisture sensitive. Store under inert gas. Store away from oxidizing agent, air, water/ moisture.
Specifications
| Tri-n-butyl phosphite | |
| -80°C | |
| 125°C to 128°C (10 mmHg) | |
| C12H27O3P | |
| MFCD00009437 | |
| 1703866 | |
| tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri | |
| XTTGYFREQJCEML-UHFFFAOYSA-N | |
| tributyl phosphite | |
| 7623 | |
| 94% |
| 102-85-2 | |
| 0.915 | |
| 82°C (179°F) | |
| 1.432 | |
| 500 mL | |
| Air and moisture sensitive | |
| Not miscible or difficult to mix in water. | |
| CCCCOP(OCCCC)OCCCC | |
| 250.319 | |
| 250.32 |
Chemical Identifiers
| 102-85-2 | |
| 250.319 | |
| XTTGYFREQJCEML-UHFFFAOYSA-N | |
| 7623 | |
| CCCCOP(OCCCC)OCCCC |
| C12H27O3P | |
| MFCD00009437 | |
| tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri | |
| tributyl phosphite |
Safety and Handling
GHS H Statement
H312-H315-H319-H227
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H312+H332-H315-H319
EINECSNumber : 203-061-3
RTECSNumber : TH0875000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only