missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl phosphite, 94%

Catalog No. AAL14307AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14307AE 100 mL
AAL14307AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14307AE Supplier Thermo Scientific Chemicals Supplier No. L14307AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 102-85-2 | C12H27O3P | 250.319 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.

Solubility
Not miscible or difficult to mix in water.

Notes
Air and moisture sensitive. Store under inert gas. Store away from oxidizing agent, air, water/ moisture.

Chemical Identifiers

CAS 102-85-2
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.319
MDL Number MFCD00009437
InChI Key XTTGYFREQJCEML-UHFFFAOYSA-N
Synonym tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri
PubChem CID 7623
IUPAC Name tributyl phosphite
SMILES CCCCOP(OCCCC)OCCCC

Specifications

Melting Point -80°C
Density 0.915
Boiling Point 125°C to 128°C (10 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.432
Quantity 100 mL
Beilstein 1703866
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 250.32
Percent Purity 94%
Chemical Name or Material Tri-n-butyl phosphite
Show More Show Less
Hazard Category H227-H312+H332-H315-H319
Hazard Statement GHS H Statement
H312-H315-H319-H227
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-061-3
RTECSNumber TH0875000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only