missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-butyl borate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | <-70°C |
| Density | 0.853 |
| Boiling Point | 230°C to 235°C |
| Flash Point | 93°C (199°F) |
| Refractive Index | 1.4095 |
| Quantity | 100 mL |
| Beilstein | 1703865 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 230.16 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 211-706-5 |
| RTECSNumber | ED4900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only