missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl(3-methyl-2-butenyl)tin, 95%

CAS: 53911-92-5 | C17H36Sn | 359.185 g/mol

Supplier:  Thermo Scientific Chemicals L2021406

 View more versions of this product

Catalog No. AAL2021406

Only null left
Explore available promotions

Description

Description

Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Tri-n-butyl(3-methyl-2-butenyl)tin
1.042
>110°C (230°F)
1.488
5 g
3588742
Immiscible with water.
CCCC[Sn](CCCC)(CCCC)CC=C(C)C
359.185
359.19
53911-92-5
283°C
C17H36Sn
MFCD01863649
UN2788
tributyl 3-methylbut-2-en-1-yl stannane, tributyl 3-methyl-2-butenyl tin, tributylprenylstannane, acmc-1am9p, tri-n-butyl 3-methyl-2-butenyl tin, tributyl 3-methylbut-2-enyl stannane, stannane, tributyl 3-methyl-2-butenyl, tributyl 3-methylbut-2-en-1-yl ;stannane
XEFRYQPTIWSVGI-UHFFFAOYSA-N
tributyl(3-methylbut-2-enyl)stannane
2734722
95%

Chemical Identifiers

53911-92-5
359.185
XEFRYQPTIWSVGI-UHFFFAOYSA-N
2734722
CCCC[Sn](CCCC)(CCCC)CC=C(C)C
C17H36Sn
MFCD01863649
tributyl 3-methylbut-2-en-1-yl stannane, tributyl 3-methyl-2-butenyl tin, tributylprenylstannane, acmc-1am9p, tri-n-butyl 3-methyl-2-butenyl tin, tributyl 3-methylbut-2-enyl stannane, stannane, tributyl 3-methyl-2-butenyl, tributyl 3-methylbut-2-en-1-yl ;stannane
tributyl(3-methylbut-2-enyl)stannane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only