missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl(1-ethoxyvinyl)tin, 95%

Catalog No. AAH3156206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3156206 5 g
AAH3156203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH3156206 Supplier Thermo Scientific Chemicals Supplier No. H3156206
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 97674-02-7 | C16H34OSn | 361.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97674-02-7
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00010240
InChI Key HGXJOXHYPGNVNK-UHFFFAOYSA-N
Synonym tributyl 1-ethoxyvinyl stannane, tributyl 1-ethoxyvinyl tin, tributyl 1-ethoxyethenyl stannane, 1-ethoxyvinyltributyltin, 1-ethoxyvinyl tributyltin, 1-ethoxyvinyltri-n-butyltin, 1-ethoxyvinyl tributylstannane, 1-ethoxyethenyl tributylstannane, tributyl-1-ethoxyvinyl tin, stannane, tributyl 1-ethoxyethenyl
PubChem CID 619414
IUPAC Name tributyl(1-ethoxyethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C(=C)OCC

Specifications

Density 1.067
Boiling Point 85°C to 86°C (0.07 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.476
Quantity 5 g
UN Number UN2788
Formula Weight 361.14
Chemical Name or Material Tri-n-butyl(1-ethoxyvinyl)tin
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only