missing translation for 'onlineSavingsMsg'
Learn More

Tri(cyclohexa-2,5-dien-1-yl)silane 93.0+%, TCI America™

Catalog No. T3443200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3443200MG 200 mg
T34431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3443200MG Supplier TCI America Supplier No. T3443200MG
Only null left

Chemical Identifiers

CAS 1807776-35-7
Molecular Formula C18H22Si

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 200 mg
Formula Weight 266.46
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri(cyclohexa-2,5-dien-1-yl)silane