missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Tray RTU Format Velocity™ Vials

Catalog No. 01677100
Change view
Click to view available options
Product Line:
10R
20R
2R
6R
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
01677100 20R
01677097 2R
01677098 6R
01677099 10R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01677100 Supplier Corning™ Supplier No. 91395E5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Velocity™ vials in Tray RTU format.

Corning™ Velocity™ vials

  • Type I externally coated borosilicate glass vials
  • RTU with SG EZ-fill™ Technology
  • Tray RTU format

Specifications

Material Borosilicate Glass
Volume (Metric) Brim 26 mL
Product Line 20R
Height (Metric) 55 mm
Product Type Pharmaceutical Vial