missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Tray RTU Format Type I Glass Vials

Catalog No. 01677083
Change view
Click to view available options
Product Line:
10R
20R
2R
30R
4R
6R
8R
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
01677083 2R
01677084 4R
01677085 6R
01677086 8R
01677087 10R
01677088 20R
01677089 30R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01677083 Supplier Corning™ Supplier No. 81088E5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality Type I glass vials in Tray RTU format.

  • Type I uncoated borosilicate glass vials
  • RTU with SG EZ-fill™ Technology
  • Tray RTU format

Specifications

Material Borosilicate Glass
Volume (Metric) Brim 4 mL
Product Line 2R
Height (Metric) 35 mm
Product Type Pharmaceutical Vial