missing translation for 'onlineSavingsMsg'
Learn More

Traumatic Acid 90.0+%, TCI America™

Catalog No. D0029100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0029100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0029100MG Supplier TCI America Supplier No. D0029100MG
Only null left

Chemical Identifiers

CAS 6402-36-4
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD00004443
InChI Key MAZWDMBCPDUFDJ-VQHVLOKHSA-N
Synonym trans-2-Dodecene-1,12-dioic Acid
PubChem CID 5283028
ChEBI CHEBI:545687
IUPAC Name (2E)-dodec-2-enedioic acid
SMILES OC(=O)CCCCCCCC\C=C\C(O)=O

Specifications

Melting Point 165°C
Quantity 100 mg
Formula Weight 228.29
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Traumatic Acid