Learn More
Innovatek Transwab™ Liquid Culture Swabs
Catalog No. 29801024
Change view
Click to view available options
Swab Configuration:
Dual Applicator
Single Applicator
Media Type:
Amies Charcoal
Amies Clear
Shaft Material:
Flexible Wire
Plastic
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Swab Configuration Media Type Shaft Material
29801024 Single Applicator Amies Clear Plastic
361045657 Dual Applicator Amies Clear Plastic
361030259 Single Applicator Amies Charcoal Flexible Wire
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 29801024 Supplier Innovatek Supplier No. 4080MW170
Transwab™ Liquid Culture Swabs provide superior recovery of pathogens in a variety of applications
  • Swab and labeled tube in peel pouch
  • Semi-solid or liquid transport media for superior recovery of pathogens
  • For use in medical, clinical, food, forensic, environmental, and industrial applications
  • Choose tip material and size, shaft material, liquid or gel transport media to suit every need
Specifications
Media Type Amies Clear
Swab Configuration Single Applicator
For Use With (Application) Wound, Skin, Urogenital, and Throat Swabs
Quantity 125 Swabs/Box
Shaft Material Plastic
Tip Material Rayon