GA International Transparent Removable Cryogenic Labels for Laser Printers 2"" x 1.125"" / 50.8mm x 28.6mm Rectangle Transparent From -196C to +120C (-321F to +248F)
Catalog No. 5000571796
Transparent cryogenic thermoplastic laser labels with a glove friendly removable adhesive. Ideal for long-term storage identification of flat-surface containers in liquid nitrogen and harsh deep-freeze environments used in biomedical research and clinical laboratories. The clear cryo labels accept desktop laser printing and permanent marker. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.
Catalog No.
5000571796
Supplier
GA InternationalSupplier No.
TRCL63
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Transparent cryogenic thermoplastic laser labels with a glove friendly removable adhesive. Ideal for long-term storage identification of flat-surface containers in liquid nitrogen and harsh deep-freeze environments used in biomedical research and clinical laboratories. The clear cryo labels accept desktop laser printing and permanent marker. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.