missing translation for 'onlineSavingsMsg'
Learn More

Transparent Lifting Tape

Catalog No. S99535
Change view
Click to view available options
Roll size: 5cm x 9.1m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S99535 Roll size: 5cm x 9.1m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S99535 Supplier Customer Channels Group (Ccg) Supplier No. 144L2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conforms to most surfaces.

  • Pressure wound, heat treated and “fish-eye” free
  • After lifting powdered prints, tape easily mounts on rigid vinyl or coated paper backing sheets
  • Supplied in a dust-free polystyrene jar
  • Roll size: 2 in. × 30 ft. (5cm × 9.1m)
  • Wound on standard 1 in. core to fit lifting tape dispensers

Specifications

Color Transparent
Includes Dust-free polystyrene jar
Product Type Lifting Tape