missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio TransIT-AAViator Transfection System

Catalog No. MIR73750
Catalog No. MIR73750 Supplier Mirus Bio Supplier No. MIR 73750
Only null left

A new transfection system tailored specifically for maximal AAV production.

Developed on a proven polymer and lipid technology platform that provides:

  • Increased AAV vector output
  • Easier scalability
  • Reduced plasmid DNA consumption by up to 50% compared to Polymer-only reagents.

The TransIT-AAViator System is made up of the new TransIT-AAViator Transfection Reagent and RevIT™ AAV Enhancer. When combined, process developers now have a transfection system tailored specifically for maximal AAV production.

Specifications

Cell Type HEK293, 293-derived Cells
Format Kit
Promoter None (Promoterless)
Transfection Technique Lipid-based Transfection, Transient Transfection
Product Type Transfection System
Sterility Sterile-filtered