Learn More
XpressBio TRANSIL PPB BINDING KIT
Catalog No. NC2333892
Encompass
The Sovicell TRANSIL PPB Binding Kit estimates the binding of drugs to human serum albumin (HSA) and human alpha-1 glycoprotein (AGP/AAG) at a physiological ratio to predict plasma protein binding. Internal quality controls provide easy assessment of recovery, experiment and data quality.
Catalog No. NC2333892 Supplier XpressBio Supplier No. TPB02122096
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Sovicell TRANSIL PPB Binding Kit estimates the binding of drugs to human serum albumin (HSA) and human alpha-1 glycoprotein (AGP/AAG) at a physiological ratio to predict plasma protein binding. Internal quality controls provide easy assessment of recovery, experiment and data quality.