These pipets are good for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The non-sterile pipets are bulk packed; the sterile are individually packed.
Bulb Draw: 4.6 ml
Drops/mL: 20
OAL: 15.7 cm
Stem diameter: 6.1 cm
Package: Bulk