missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transformation buffer-2, pH 7.5, Thermo Scientific™
For transformation of competent cells
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63293EQE
|
Thermo Scientific Chemicals
J63293EQE |
125mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AAJ63293AK
|
Thermo Scientific Chemicals
J63293AK |
250mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsTransformation buffer-2 is used for transformation of competent cells.
Solubility
Fully miscible in water.
Notes
Contains 10mM MOPS, 75mM calcium chloride, 10mM potassium chloride, and 15% glycerol, pH 7.5
Specifications
| 7.5 |