missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Transferpette™ Electronic Mount

Catalog No. 14381081
Change view
Click to view available options
Transferpette™ Electronic Mount
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14381081 Transferpette™ Electronic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14381081 Supplier BRANDTECH™ BRAND™ Supplier No. 705383

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accessory for use with the Transferpette electronic single and multichannel pipettes.

  • Allows hanging of pipette on shelf, or as a replacement part for benchtop rack
  • Pipettes sold separately

Specifications

Color Black
For Use With BRAND Transferpette Electronic
Material Polycarbonate
Model Shelf/rack mount for Transferpette electronic
Number of Channels 1 or 8
For Use With (Equipment) Shelf/Rack Mount