Learn More
Synthware™ Transfer Valve
The 0 to 4 mm high vacuum valves have two Viton O-rings on the shaft and an integral PTFE O-ring on the tip. The standard taper upper outer joint and lower inner joint are the same size.
$326.11 - $334.54
Specifications
| For Use With (Application) | Organic Synthesis |
|---|---|
| Material | Borosilicate Glass |
| Quantity | 1 Ea. |
| Width (Metric) | 75 mm |