missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Transfer Rack for Eppendorf SmartBlock™

Catalog No. 05414273
Change view
Click to view available options
Holds:
24 x 1.5/2.0 mL
24 x 0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
05414273 24 x 0.5 mL
05414272 24 x 1.5/2.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05414273 Supplier Eppendorf™ Supplier No. 3880000305

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Transfer rack suitable for individually sensor-controlled SmartBlock™ heating/cooling/mixing blocks.

  • Holds up to 24 microtubes for easy transfer of multiple vessels in parallel.
  • Convenient transfer of up to 24 hot vessels without burning the fingers
  • Temperature range from -86°C to 121°C for broad flexibility
  • Limited to ThermoMixer C annd ThermoStat C

Specifications

Holds 24 x 0.5 mL
For Use With (Equipment) SmartBlock, ThermoMixer
Temperature (Metric) Range −86°C to 121 °C