Tradewinds Direct Inc Transfer Pipette, Twdbiox, 1Ml, Disposable, Non-Sterile, Polyethylene, Molded Graduations, 2000/Case.
Catalog No. 502285026
1Ml Disposable Polyethylene Non-Sterile General Purpose Transfer Pippets (Length: 160Mm | Capacity: 6.2Ml). Pippets Have Molded Graduations At Each 0.25Ml Up To 1Ml (26 Drops/Ml). 500 Pipettes/Pack, 4 Packs/Case, Price/Case
Catalog No.
502285026
Supplier
Tradewinds Direct IncSupplier No.
BX3001351
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
1Ml Disposable Polyethylene Non-Sterile General Purpose Transfer Pippets (Length: 160Mm | Capacity: 6.2Ml). Pippets Have Molded Graduations At Each 0.25Ml Up To 1Ml (26 Drops/Ml). 500 Pipettes/Pack, 4 Packs/Case, Price/Case