Tradewinds Direct Inc Transfer Pipette, Twdbiox, 3Ml, Disposable, Sterile, Polyethylene, Molded Graduations, Individually Wrapped, 2000/Case.
Catalog No. 502285030
3Ml Disposable Polyethylene Sterile General Purpose Transfer Pippets (Length: 184Mm | Capacity: 7.5Ml). Pippets Have Molded Graduations At Each 0.5Ml Up To 3Ml (22 Drops/Ml). Pippets Come Individually Wrapped. 500 Pipettes/Pack, 4 Packs/Case, Price/Case
Catalog No.
502285030
Supplier
Tradewinds Direct IncSupplier No.
BX300238A1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
3Ml Disposable Polyethylene Sterile General Purpose Transfer Pippets (Length: 184Mm | Capacity: 7.5Ml). Pippets Have Molded Graduations At Each 0.5Ml Up To 3Ml (22 Drops/Ml). Pippets Come Individually Wrapped. 500 Pipettes/Pack, 4 Packs/Case, Price/Case