Learn More
Tradewinds Direct Inc Transfer Pipette, Twdbiox, 3Ml, Disposable, Non-Sterile, Polyethylene, Molded Graduations, 2000/Case.
Catalog No. 502285028
Encompass
3Ml Disposable Polyethylene Non-Sterile General Purpose Transfer Pippets (Length: 160Mm | Capacity: 7.5Ml). Pippets Have Molded Graduations At Each 0.25Ml Up To 3Ml (22 Drops/Ml). 500 Pipettes/Pack, 4 Packs/Case, Price/Case
Catalog No. 502285028 Supplier Tradewinds Direct Inc Supplier No. BX3001381
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
3Ml Disposable Polyethylene Non-Sterile General Purpose Transfer Pippets (Length: 160Mm | Capacity: 7.5Ml). Pippets Have Molded Graduations At Each 0.25Ml Up To 3Ml (22 Drops/Ml). 500 Pipettes/Pack, 4 Packs/Case, Price/Case