Learn More
Research Products Corporation Disposable Plastic Transfer Pipettes, Sterile, Graduated Fine Tip, 3.9ml Capacity, Individually Wrapped, 500 per package
Catalog No. 501367947
Encompass
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling.
Catalog No. 501367947 Supplier Research Products Corporation Supplier No. 1475071S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling.