missing translation for 'onlineSavingsMsg'
Learn More

Azure Biosystems Transfer Buffer

Catalog No. AC2127AZ
Catalog No. AC2127AZ Supplier Azure Biosystems Supplier No. AC2127

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Transfer buffer, 50x concentrate, sufficient for 50 to 100 blots

  • This proprietary transfer buffer provides improved detection of low-abundance and post-translationally modified proteins.
  • High-efficiency protein transfer and increased protein retention on the membrane add up to more sensitive Western blots.
  • Provided as a 50x concentrated solution. Sufficient for 50 to 100 blots
  • Size: 500 mL

Specifications

Chemical Name or Material Transfer Buffer
Concentration 50X
For Use With (Application) Western Blot
Physical Form Solution