missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Transfer Buffer Powder For Use with mPAGE™ Bis-Tris Gels
Description
- Transfer buffer powder for use with mPAGE Bis-Tris gels.
- 10 packets. Each packets makes 1 liter of transfer buffer at 1X.
- Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Specifications
Specifications
| Chemical Name or Material | Transfer buffer powder |
| Physical Form | Powder |
| Quantity | 10 Pk. |