missing translation for 'onlineSavingsMsg'
Learn More

trans-Urocanic Acid 98.0+%, TCI America™

Catalog No. I00025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I00025G Supplier TCI America Supplier No. I00025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3465-72-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00005203,MFCD01593677
InChI Key LOIYMIARKYCTBW-OWOJBTEDSA-N
Synonym 4-Imidazoleacrylic Acid
PubChem CID 736715
ChEBI CHEBI:30817
IUPAC Name (2E)-3-(1H-imidazol-5-yl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CN=CN1

Specifications

Melting Point 225°C
Color White
Quantity 5 g
Formula Weight 138.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-Urocanic Acid
EINECSNumber (5)-3517
RTECSNumber NI3425200
TSCA Yes
Recommended Storage Refrigerator